Order Entry
ContactUsLinkComponent
TUBESTORAGE60MLVALVECAJON
Vacuum Storage Tube, with Hi-Vac TopValve and ⅜" Cajon Sidearm, Ace Glass Incorporated
Catalog #: 89427-978
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
TUBESTORAGE60MLVALVECAJON
Vacuum Storage Tube, with Hi-Vac TopValve and ⅜" Cajon Sidearm, Ace Glass Incorporated
Catalog #: 89427-978
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Capacity:
    60 ml
  • Description:
    Vacuum Storage Tube, with Hi-Vac Top Valve and 3/8" Cajon Sidearm
  • Cat. no.:
    89427-978
  • Supplier No.:
    7813-07

Specifications

About this item

This borosilicate storage tube is supplied with an 0-4mm Hi-Vac valve top with a 9mm side port designed to accept ⅜" Cajon fittings

The valved top enables precise flow control.

Customers Who Bought This Also Bought