Filtration Apparatus, Ace Glass Incorporated
Catalog #: 89427-882
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Membrane Filtration Apparatus, 75 mm, Includes 2000 mL Filter Flask with Ace-Safe Connection,1000 mL Graduated Funnel, Porosity C (25-50 micron) Fritted Disc, PTFE Adapter, and Retaining Ring
- Cat. no.:89427-882
- Supplier No.:3708-02
Specifications
About this item
This borosilicate glass filtration apparatus accomodates 75 mm membrane filters.
The new design utilizes a PTFE Ace-Thred adapter to join the glass funnel, filter flask and fritted disc together. This design eliminates the need for the clamping device, stopper and base. The unit is easy to assemble, disassemble and clean. The fritted disc can be easily replaced and other porosity discs used. The apparatus is easier to use and more economical than old designs.
Customers Who Bought This Also Bought