OdorGone® Autoclave Deodorant Capsules, DLI, Decon Labs
Catalog #: CA11027-240
Supplier: Decon Labs
Specifications
- Description:Lemon Scented
- Cat. no.:CA11027-240
- Supplier No.:LEM100
Specifications
About this item
OdorGone® Autoclave Deodorant Capsules are activated by the heat and moisture from the autoclave to release the contents of the capsule and overcome unpleasant smells generated from heating microbiological waste
One capsule per load absorbs offensive odors leaving the laboratory smelling fresh and clean. Available in apple or lemon frangrance.
Certifications: UL listed.
Customers Who Bought This Also Bought