Order Entry
ContactUsLinkComponent
VIALMICROWAVE2MLCONICAL BTM
Microwave Reaction Vials, Ace Glass Incorporated
Catalog #: 89427-726
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
VIALMICROWAVE2MLCONICAL BTM
Microwave Reaction Vials, Ace Glass Incorporated
Catalog #: 89427-726
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Cap included:
    No
  • Capacity:
    0.20 - 2.00 ml
  • Description:
    Microwave reaction vials
  • Cat. no.:
    89427-726
  • Bottom Style:
    Conical
  • Supplier No.:
    5704-02

Specifications

About this item

Heavy wall, Type 1 clear borosilicate glass for higher pressure, microwave digestion.

20 mm flange top to accomodate crimp seals. Available with either conical or rounded bottoms.

Customers Who Bought This Also Bought