Hi-Recovery Vials, Ace Glass Incorporated
Catalog #: 89427-720
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Cap included:No
- Cap Size:15-425
- Capacity:5.00 ml
- Description:Hi-Recovery Vial, Tapered Bottom
- Cat. no.:89427-720
- Supplier No.:5703-05
Specifications
About this item
Clear type 1 borosilicate glass vials with recessed bottom for both concentration of material during reactions, extractions and chromatography applications.
Vials are either straight sided with a conical bottom or a tapered internal bottom.
Customers Who Bought This Also Bought