Soxhlet Extractor, Glassco
Catalog #: 89426-700
Supplier: Glassco
Specifications
- Description:Soxhlet Extractor
- Cat. no.:89426-700
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 34/45
- Supplier No.:210.470.01
Specifications
About this item
Extractor body only.
Customers Who Bought This Also Bought