Round Bottom Flask, Single Neck, Glassco
Catalog #: 89426-460
Supplier: Glassco
Specifications
- Capacity:100 ml
- ST Joint No.:24/40
- Description:Round Bottom Flask, Single Neck
- Supplier No.:057.202.58
Specifications
About this item
Heavy wall design with single neck
Made from Borosilicate 3.3 glass. All products listed are hand blown from tubing to ensure uniform thickness.
Customers Who Bought This Also Bought