Anti-Splash Adapter, Glassco
Catalog #: 89426-396
Supplier: Glassco
Specifications
- Capacity:100 ml
- Description:Anti-Splash Adapter
- Cat. no.:89426-396
- Bottom [ST] Joint Size:14/20
- Top ST Joint Size:[ST] 24/40
- Supplier No.:030.475.04
Specifications
About this item
Anti-Splash adapters are used between the vapor tube and evaporation flask on a rotary evaporator to prevent the contents of the flask from being drawn into the condenser.
- Sealed inner vapor tube
- Modified bump traps for expanded volume
Modified bump traps provide expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the trap to prevent entry into the condenser.
Customers Who Bought This Also Bought