Enlarging Adapter, Glassco
Catalog #: 89426-336
Supplier: Glassco
Specifications
- Description:Enlarging Adapter
- Cat. no.:89426-336
- Bottom [ST] Joint Size:14/20
- Top ST Joint Size:[ST] 24/40
- Supplier No.:014.470.02
Specifications
About this item
Enlarging adapters are useful for attaching apparatus with one joint size to equipment having a larger joint size.
- Top [ST] joint
- Bottom [ST] joint
Enlarging adapters are equipped with joints at top and bottom.
Customers Who Bought This Also Bought