Bushing Adapter, Glassco
Catalog #: 89426-322
Supplier: Glassco
Specifications
- Description:Bushing Adapter
- Cat. no.:89426-322
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 14/35
- Supplier No.:013.478.02
Specifications
About this item
Bushing adapters allow for compact assemblies of dissimilar joints.
- Top [ST] joint
- Bottom [ST] joint
- Manufactured from borosilicate 3.3 glass
Bushing-style adapters have upper and lower standard taper joints.
Customers Who Bought This Also Bought