Dropette® Disposable Transfer Pipettes, Simport Scientific
Catalog #: CA11027-120
Supplier: Simport Scientific
Specifications
- Capacity:3 ml
- Material:Polyethylene
- Cat. no.:CA11027-120
- Overall Length:13.8 cm
- Bulb Draw:2.1 ml
- Supplier No.:P200-30
Specifications
About this item
These all-in-one pipettes eliminate the hazard of broken glass and exposure to infectious materials.
- Can be sealed and refrigerated
- Temperature resistant down to –196 °C
- Can be gas (EtO) sterilized
- Non-toxic and inert
- No bulb to insert or remove
- Uniform drop size
Put an end to matching rubber bulbs with glass pipettes. Molded from see-through low density polyethylene. Inert to biological fluids and most acids. The low-affinity surface reduces the loss of cells and valuable proteins due to binding.
Customers Who Bought This Also Bought