To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Ø:12 mm (16 mm Cap Dia.)
- Length:120 mm
- Description:Combination flat surface pH electrode, with BNC connector
- Operating temperature range:0 to 100°C
- Body material:Epoxy
- Cat. no.:34107-650
- Cable Length:1 m
- Supplier No.:8135BN
Specifications
About this item
Stable readings to 0.01 pH in less than 30 s.
- pH range of 0 to 14
- Glass fiber junction
- pH precision: 0.01
The Orion™ ROSS™ Combination Flat Surface pH Electrode measures pH of soft, moist surfaces; both solid and semi-solid. This includes agar gel plates, meats, bread dough, and similar samples. With drift less than 0.002 pH per day and minimized restandardization, this electrode works in extreme cases of samples varying from one another by 50 °C or more. Includes one bottle of 3 M KCl filling solution.
Get results three to five times more precise than those obtained with conventional electrodes with the Orion™ ROSS™ Combination Flat Surface pH Electrode.