Disodium hydrogen phosphate dihydrate ≥99.5% (by acidimetry), EMSURE® for analysis, Supelco®
Catalog #: CA1.06580.0500
Supplier: MilliporeSigma
CAS Number: 10028-24-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic bottle
- Size:500 g
- Cat. no.:CA1.06580.0500
- Supplier No.:1.06580.0500
Specifications
Specification Test Results
- Assay (acidimetric):≥ 99.5 %
- Identity:passes test
- pH-value (5 %; water):9.0 - 9.2
- Chloride (Cl):≤ 0.001 %
- Sulfate (SO₄):≤ 0.005 %
- Total nitrogen (N):≤ 0.001 %
- Heavy metals (as Pb):≤ 0.001 %
- Cu (Copper):≤ 0.0003 %
- Fe (Iron):≤ 0.001 %
- K (Potassium):≤ 0.005 %
- Pb (Lead):≤ 0.001 %
- Supelco®, Disodium hydrogen phosphate dihydrate ≥99.5% (by acidimetry), EMSURE® for analysis:
Specification Test Results
About this item
Synonyms: sec-Sodium phosphate,Sodium monohydrogen phosphate dihydrate,Sodium hydrogenphosphate dihydrate,di-Sodium hydrogen orthophosphate dihydrate,Sodium phosphate dibasic dihydrate,Disodium phosphate,di-Sodium hydrogen phosphate dihydrate
Melting Pt:92.5 °C (–H₂O of crystall.)
Density:2.1 g/cm3 (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 92.5 °C (–H₂O of crystall.)
:Flash Pt:
:Flash Pt:
:M.W. 177.99 g/mol
EINECS:231-448-7
:92.5
Storage Temperature:Ambient
UN:3077
Customers Who Bought This Also Bought