Ward's® Physiology of the Circulatory System Lab Activity
Catalog #: 470194-706
Supplier: Ward's Science
Specifications
- Description:Live Daphnia magna, 100+ specimens, for AP Biology Lab #10
- No. of Lab Groups:—
- Cat. no.:470194-706
- Supplier No.:WARD470194-706
Specifications
About this item
View Daphnia Heart Rate Easily
- Teaches technique for measuring blood pressure
- Comparative physiology featuring Daphnia
- Specifically designed viewing chambers for viewing Daphnia
- Time Required: 75 minutes
Students learn how to measure blood pressure, pulse, and heart rate, and assess the effects that varying physiological conditions have on their rates, in this practical, comprehensive lab activity. For comparison, students also study the heart rate of Daphnia, varying the temperature and viewing the subsequent changes that take place in the ectothermic creature. Ward's has included the components of a viewing chamber that students make themselves for the easiest viewing of the Daphnia specimens.
Materials required, not included: sphygmomanometer (blood pressure cuff)
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Customers Who Bought This Also Bought