Coplin Staining Jars
Catalog #: 470174-652
Supplier: United Scientific Supplies
Specifications
- Capacity:Up to 10 Slides
- Description:Coplin Staining Jar, Plastic
- Cat. no.:470174-652
- Supplier No.:62101-A
Specifications
About this item
Coplin Jar with Screw-on Top Prevents Spills.
- Quality slide making supplies
- Slide staining tools
The screw-on cover prevents evaporation and contamination when staining. Each jar holds five single slides or ten back to back.
Customers Who Bought This Also Bought