Vermiculite
Catalog #: 470021-148
Supplier: Avantor
Specifications
- Description:Vermiculite, 4 qts.
- Cat. no.:470021-148
- Supplier No.:WARD470021-148
Specifications
About this item
Light, porous mica medium retains moisture well, making it excellent for starting seeds or as a soil additive.
Customers Who Bought This Also Bought