Herbarium Paste
Catalog #: 470021-054
Supplier: Avantor
Specifications
- Description:Herbarium Paste
- Cat. no.:470021-054
- Supplier No.:WARD470021-054
Specifications
About this item
Specially formulated to adhere plant specimens to mounting sheets.
- Waterbased
- Easy cleanup
- Low odor
This longtime industry standard paste is waterbased for easy cleanup and low odor. The dispenser tip measures just the right amount of paste to attach herbarium specimens to mounting sheets. One pint bottle.
Customers Who Bought This Also Bought