Ward's® Microhematocrit Set
Catalog #: 470019-794
Supplier: Avantor
Specifications
- Description:WARD’S Microhematocrit Set
- Cat. no.:470019-794
- Supplier No.:WARD470019-794
Specifications
About this item
The set includes 200 heparinized, mylar wrapped, hematocrit tubes, and six capillary tube trays with sealant. Each tray has two rows of slots numbered 1 through 24 for easy tube storage.
Customers Who Bought This Also Bought