Chemical Storage Start-Up Kit, Ward's
Catalog #: 470010-858
Supplier: Avantor
Specifications
- Description:Ward´s® chemical storage start-up kit
- Cat. no.:470010-858
- Supplier No.:WARD470010-858
Specifications
About this item
This kit maintain a safe environment by storing chemicals properly.
- Detailed instructions
- Chemical storage posters
- Color-coded system
This kit helps you create a consistent storage classification system. The provided color-coding system allows you to label chemicals for correct use and storage and by compatibility, so you have an effective storage system that’s compliant with industry standards.
For each of the five colors used in this system (green, white, yellow, red, and blue), there are enough label dots included to re-label 350 bottles and sixty-nine 3' shelves.
Certifications: OSHA/WHMIS-Compliant.
Ordering information: This kit includes rolls of tape and label dots, detailed instructions, one large Ward's chemical storage system poster, and four small posters ¹/₂.
Customers Who Bought This Also Bought