Giant Water Bug
Catalog #: 470001-304
Supplier: Avantor
Specifications
- Description:Giant Water Bug, Preserved, Jar of 10
- Cat. no.:470001-304
- Supplier No.:WARD470001-304
Specifications
About this item
Alcohol Preserved
Lethocerus or Belastoma. Lengthened, flattened hind legs serve as oar blades, propelling the insect through the water. Size: 1".
Customers Who Bought This Also Bought