Laminaria
Catalog #: 470001-168
Supplier: Avantor
Specifications
- Description:Laminaria, Preserved, Jar, Each
- Cat. no.:470001-168
- Supplier No.:WARD470001-168
Specifications
About this item
Formalin Preserved
Kelp. Single complete specimen. Includes holdfast.
Supplied in a jar of 50% glycerine. Includes enough materials for 10 students, unless specified as a jar of each.
Customers Who Bought This Also Bought