Ward's® Special Sand-Gravel Mix and Stress Clay
Catalog #: 470029-384
Supplier: Avantor
Specifications
- Description:Stress Clay, 20 lbs.
- Cat. no.:470029-384
- Supplier No.:WARD470029-384
Specifications
About this item
Specially prepared sediment mixes for use with stream tables.
- Created by Ward's geologists
- Compatible with ay type of stream table
- Teach students about stress, fractures, and faults with real earth materials.
Sand-gravel mix includes various sediments up to ¹/₈" diameter. Stress clay is perfect for showing the development of stress cracks. Both mixes come packaged in reusable pails.
Customers Who Bought This Also Bought