Specifications
- Description:WARD’S Bulk Simulated Antisera, Anti-B, 1 L
- Cat. no.:470024-648
- Supplier No.:WARD470024-648
Specifications
About this item
At the request of our college customers, WARDS now offers these convenient bulk refills for our exclusive simulated ABO and Rh Blood-Typing Lab Activity (36 V 0019).
Instructions for performing blood typing using WARDS Simulated Blood and Simulated Antisera can be found in the teacher’s guide and student copymaster that accompanied the purchase of the lab activity. Each antiserum is provided in a one liter bottle.