Order Entry
ContactUsLinkComponent
INVESTIGATING DIGESTIVE PROCESSES LAB
Ward's® Investigating Digestive Processes Kit
Catalog #: 470015-800
Supplier:  Avantor
INVESTIGATING DIGESTIVE PROCESSES LAB
Ward's® Investigating Digestive Processes Kit
Catalog #: 470015-800
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Investigating Digestive Processes Kit
  • No. of Lab Groups:
    6
  • Cat. no.:
    470015-800
  • Supplier No.:
    WARD470015-800

Specifications

About this item

With this kit, students discover how our bodies break down food.

  • Students research how bodies process food by recreating digestive system
  • Activities include altering enzymes, testing for digestion byproducts, and more
  • Pre- and post-laboratory assessments included
  • Material for six setups
  • Estimated lab time–day one: 60 minutes, day two: 15 minutes
  • Aligned to NGSS standards (MS/HS: PS3, LS1)

Before food can be used as fuel, certain enzymes in various parts of the body must break it down into a usable form. Students can re-create this process and observe the action of different types of enzymes found in the body.

They can alter pH and temperature to see how these factors affect the enzymes, determine where in the body starches, proteins, and fats are digested, and test for digestion byproducts. The kit includes a teacher’s guide, a student copymaster, and material for six setups.

The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.

Ordering information: This kit includes pipets, pH test strips, indicator charts, iodine, olive oil, hydrochloric acid (1%), sodium hydroxide (0.1%), starch solution (1%), pancreatin, pepsin to make 1% solution, phenolphthalein solution, and test tubes with screwcaps. Hard-boiled eggs (one per group), an incubator, an ice bath, and test tube racks are required but not included.
Customers Who Bought This Also Bought