Ward's® Instant Drosophila Medium
Catalog #: 470015-394
Supplier: Avantor
Specifications
- Description:WARD’S Instant Drosophila Medium, Blue, 1 L
- Cat. no.:470015-394
- Supplier No.:WARD470015-394
Specifications
About this item
Improved Formula for Higher Yields
This ready-to-use formula, developed by WARD’S, just needs water. No cooking, sterilization, or additional inhibitors are necessary to create the perfect environment for culturing Drosophila. In addition, the medium won’t soften or liquefy, so flies won’t drown. Choose white or blue for better observation of developing larvae. The color won’t harm your Drosophila. One liter is sufficient for approximately 60 standard cultures.
Customers Who Bought This Also Bought