Specifications
- Description:Ward's® simulated ABO and Rh blood-typing kit, refill kit
- No. of Lab Groups:12
- Cat. no.:470015-184
- Supplier No.:WARD470015-184
Specifications
About this item
Using Ward's safe and nontoxic simulated blood, students can simulate a blood-typing experiment and delve into the concepts of Rh typing.
- Students Employ the Concepts of Structures and Properties (LS1) and Heredity (LS3)
- Kit Aligns with National Standards
- Material for 12 Groups of Students
- Estimated Lab Time—Part A: 30 Minutes, Part B: 20 Minutes
- Assessments and Vocabulary Words Target Common Core Standards
Upon completion of this kit, students will have typed an unknown sample of blood. During this process, they employ the basic science framework concepts of Structures and Properties (LS1) and Heredity (LS3) to examine the risks of Rh incompatibilities and view simulated red and white blood cells under a microscope.
This kit aligns with all published National Standards. Pre and post-laboratory assessments and vocabulary words target specific Science and Engineering Practices and Common Core Standards.