ContactUsLinkComponent
TEST KIT WATER QUALITY 50 STRIPS
Ward's® Instant Water Quality Test Kit
Catalog #: 470014-998
Supplier:  Avantor
TEST KIT WATER QUALITY 50 STRIPS
Ward's® Instant Water Quality Test Kit
Catalog #: 470014-998
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    WARD’S Instant Water Quality Test Kit
  • Cat. no.:
    470014-998
  • Supplier No.:
    WARD470014-998
Specifications
About this item
Test Five Factors on One Test Strip
Your students will be amazed to learn that their drinking glasses may hold more than just water. Have them bring water samples to class or take the test into the field, and this water quality testing kit will do the rest. Step-by-step instructions, as well as background information on each testing parameter is included. Test for total and free chlorine, total hardness, total alkalinity, and pH simultaneously on one test strip. The kit includes 50 test strips.
Customers Who Bought This Also Bought