Northern Sea Cucumber
Catalog #: 470000-776
Supplier: Avantor
Specifications
- Description:Northern Sea Cucumber, Preserved, Pail of 10
- Size:3–4"
- Cat. no.:470000-776
- Supplier No.:WARD470000-776
Specifications
About this item
Formalin Preserved
Cucumaria. The specimen represents the Holothuroidea class, an important class of echinoderms, which display the characteristics of the phylum. With their sausage-like shape and modified pentamerous symmetry, however, they are markedly different from the sea stars and sea urchins..
Customers Who Bought This Also Bought