Ward's® Live Leeches
Catalog #: 470189-206
Supplier: Boreal Laboratories
Specifications
- Description:Live leeches
- Cat. no.:470189-206
- Packaging:Pack of 6
- Body Size:1 - 2"
- Supplier No.:WARD470189-206
Specifications
About this item
Erpobdella obscura
- Freshwater leeches
Typical examples of the class Hirudinea, these freshwater leeches are dorsoventrally flattened with prominent terminal suckers.
The species supplied may be blood suckers, predators, or scavengers.
Ordering information: These products contain living or perishable materials. Seasonal item, available from April to October, depending on weather.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought