Order Entry
ContactUsLinkComponent
STIRRING ROD GLASS 300MM L 10MM D
Glass Stirring Rods
Catalog #: 470189-016
STIRRING ROD GLASS 300MM L 10MM D
Glass Stirring Rods
Catalog #: 470189-016

Specifications

  • Ø:
    10 mm
  • Length:
    304 mm
  • Cat. no.:
    470189-016
  • Supplier No.:
    GSR012-W

Specifications

About this item

Glass stirring rod features fire polished flint glass.

  • Rounded ends

Stirring rods are made of soft flint glass, with both ends fire-polished and rounded.

Customers Who Bought This Also Bought