Ward's® Painted Lady Butterflies
Catalog #: 470188-574
Supplier: Insect Lore
Specifications
- Description:Small Butterfly Kit, 5 Larvae on Food and Habitat (Pre-mixed, moist)
- Cat. no.:470188-574
- Supplier No.:WARD470188-574
Specifications
About this item
Vanessa cardui Butterflies form chrysalides and transform into colorful butterflies right before your eyes.
- Complete habitats available in 3 different sizes
- Students experience a full life cycle
- Now available with pre-mixed or powdered food options
- Quick development - occurs between 14 to 26 days
All complete culture kits include a pop-up mesh pavilion, larvae, setup instructions, and enough media to take them from caterpillar to butterfly.
Ordering information: These products contain living or perishable materials. Please specify a delivery date at time of ordering.
Ward's holds valid USDA permits allowing us to ship to most Continental US States.
Ward's holds valid USDA permits allowing us to ship to most Continental US States.
Delivery: FOR CANADIAN CUSTOMERS–Please see the Butterfly Order Form on the Product Resources Tab for larvae spring shipping schedule.
Customers Who Bought This Also Bought