Trachea and Esophagus Slide
Catalog #: 470182-902
Supplier: Avantor
Specifications
- Description:Trachea & Esophagus Slide
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-902
- Orientation:Section
Specifications
About this item
Components of the respiratory system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Illustrates the relationship between these two structures.
Customers Who Bought This Also Bought