Digestive System Composite Slide
Catalog #: 470182-864
Supplier: Avantor
Specifications
- Description:Digestive System Composite Slide
- Cat. no.:470182-864
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470182-864
- Orientation:Section
Specifications
About this item
Compare and contrast mammalian digestive tissue
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Three regions: stomach, or fundic portion, small intestine, and large intestine.
Customers Who Bought This Also Bought