Cartilage, Composite Slide
Catalog #: 470182-642
Supplier: Avantor
Specifications
- Description:Cartilage, Composite Slide
- Cat. no.:470182-642
- Stain:Verhoeff and eosin
- Supplier No.:WARD470182-642
- Orientation:Section
Specifications
About this item
Investigate the characteristics of cartilage with and without elastic fibers
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Section of hyaline and elastic cartilage showing the presence and absence of elastic fibers.
Customers Who Bought This Also Bought