Pneumocystis carnii Slide
Catalog #: 470181-974
Supplier: Avantor
Specifications
- Description:Pneumocystis carnii
- Cat. no.:470181-974
- Stain:Periodic Acid Schiff Reaction
- Supplier No.:WARD470181-974
- Orientation:Section
Specifications
About this item
Can Cause Pneumonia in Immunocompromised Individuals
- Identifying features clearly distinguishable
- Rigorous quality control standards
In human lung. Secondary infection often observed in AIDS patients.
Customers Who Bought This Also Bought