Aurelia, Ephyra
Catalog #: 470181-538
Supplier: Avantor
Specifications
- Description:Aurelia, Ephyra
- Cat. no.:470181-538
- Stain:Schneider's and fast green
- Supplier No.:WARD470181-538
- Orientation:Whole mount
Specifications
About this item
Juvenile jellyfish
- Identifying features clearly distinguishable
- Rigorous quality control standards
Free-swimming young medusa.
Customers Who Bought This Also Bought