Lilium Ovary, Binucleate Embryo Sac Slide
Catalog #: 470181-262
Supplier: Avantor
Specifications
- Description:Lilium Ovary, Binucleate Embryo Sac Slide
- Cat. no.:470181-262
- Stain:Quadruple stained
- Supplier No.:WARD470181-262
- Orientation:Cross section
Specifications
About this item
Investigate flowering plant meiosis
- Identifying features clearly distinguishable
- Rigorous quality control standards
Lily. Completion of the first meiotic division in megaspore mother cell. Forms two free nuclei.
Customers Who Bought This Also Bought