Lycopoduim, Mature Strobilus Slide
Catalog #: 470181-144
Supplier: Avantor
Specifications
- Description:Lycopoduim, Mature Strobilus
- Cat. no.:470181-144
- Stain:Quadruple stained
- Supplier No.:WARD470181-144
- Orientation:Longitudinal section
Specifications
About this item
Flowerless Plant that Reproduces with Spores
- Identifying features clearly distinguishable
- Rigorous quality control standards
Clubmoss. A fern ally. Shows strobilus containing sporangia.
Customers Who Bought This Also Bought