Ward's® Hay Medium Solution
Catalog #: 470180-766
Supplier: Avantor
CAS Number:
Specifications
- Packaging:Bottle
- Description:Hay Medium: Concentrated
- Size:125 mL
- Cat. no.:470180-766
- Supplier No.:WARD470180-766
Specifications
About this item
Similar medium to Similar to Cereal Grass medium and Cerophyll. For culture of protists.
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Concentrate comes with a vial of heat-treated wheat seeds.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought