Ward's® Prepared Brain-Heart Infusion Agar
Catalog #: 470180-706
Supplier: IPM SCIENTIFIC, INC.
CAS Number:
Specifications
- Packaging:Plates
- Description:Prepared Brain-Heart Infusion Agar
- Cat. no.:470180-706
- Supplier No.:WARD470180-706
Specifications
About this item
Brain-Heart Infusion (BHI) Agar is a general purpose media used for the cultivation of fastidous microrganisms.
Excellent media for cultivaion of such as streptococci, pneumococci and meningococci.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought