Ward's® Live Grassfrogs (Rana pipiens)
Catalog #: 470180-602
Supplier: Boreal Laboratories
Specifications
- Description:Female Grassfrogs
- Cat. no.:470180-602
- Body Size:2 - 4"
- Supplier No.:WARD470180-602
Specifications
About this item
Grass frogs are low maintenance and seasonally available, typically from September to July.
- Great for In-Class Observation
- Low-Maintenance
- Also Known as Leopard Frogs
Keep in an aquarium or terrarium with both wet and dry areas and feed crickets and/or earthworms.
Ordering information: This kit includes living or perishable materials. They may not be available in all states. Check with your local department of agriculture.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
CANADIAN CUSTOMERS: Please note that due to Canadian import restrictions, frogs can no longer be packed with sphagnum moss.
CANADIAN CUSTOMERS: Please note that due to Canadian import restrictions, frogs can no longer be packed with sphagnum moss.
Customers Who Bought This Also Bought