Ward's® Dwarf African Frog (Hymenochirus sp)
Catalog #: 470180-584
Supplier: EVERGLADES EXOTIC PLANTS 2
Specifications
- Description:Dwarf African Frog
- Cat. no.:470180-584
- Body Size:1.0 - 1.5"
- Supplier No.:WARD470180-584
Specifications
About this item
These small aquatic amphibians breed readily in a freshwater tropical aquarium.
- Frogs are very active and excellent for classroom observation
- Easy to maintain, and do not require large aquaria
They feed on Daphnia, and small earthworms. (available separately). Specimens are approximately 1 to 1.5" in size.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought