Ward's® Live Sea Anemone Culture (Aiptasia pallida)
Catalog #: 470180-520
Supplier: Avantor
Specifications
- Description:Sea Anemone (Large)
- No. of Students:100
- Cat. no.:470180-520
- Supplier No.:WARD470180-520
Specifications
About this item
Illustrate the structure, feeding habits, responses, and reproduction of a typical Anthozoan.
It feeds on brine shrimp and other small invertebrates.
Specimens are typically absent of symbionts, if you require your specimens to have symbionts present please contact customer service.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought