Ward's® Wild Type Fruit Fly (Drosophila melanogaster)
Catalog #: 470180-442
Supplier: Avantor
Specifications
- Description:D. melanogaster culture (Large)
- No. of Students:100
- Cat. no.:470180-442
- Supplier No.:WARD470180-442
Specifications
About this item
Drosophila melanogaster, the common fruit fly, is widely used in genetic studies because they express a number of mutations such as eye, wing, and body mutations.
- Great for studying sex-linked, recessive, and dominant mutations
- Easy to maintain, with short generation times
- Excellent live food for large insects such as praying mantises, as well as small reptiles, and amphibians
Drosophila demonstrates complete metamorphosis with egg, larva, pupa, and adult stages in its life cycle.
Delivery: These products contain living or perishable materials and must ship via overnight or 2-day transit. Please specify a delivery date at checkout. Wild type exhibit normal genetic characteristics (red eyes and normal wing size). Allow cultures to come to room temperature before checking under low magnification for eggs and live larvae.
Packaging: Cultures are shipped with Drosophila medium in a shatterproof vial. The pupae will usually appear within ten days of the ship date.
Customers Who Bought This Also Bought