Ward's® Live Superworms (Zophobus morio)
Catalog #: 470180-362
Supplier: Avantor
Specifications
- Description:Superworm Larvae, Pack of 50
- Cat. no.:470180-362
- Supplier No.:WARD470180-362
Specifications
About this item
Also known as king mealworms, superworms are extremely popular as a feeder insect within the pet trade.
- Easier to digest as food source than mealworms
- Study insect defense mechanisms
They are larger in size and less chitinous than regular Tenebrio, making them easier for reptiles and amphibians to digest.
Adults also secrete an unpleasant odor when threatened, making them interesting specimens for studying insect defense mechanisms.
Ordering information: This item contains live or perishable materials.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought