Ward's® Live American Cockroaches (Periplaneta americana)
Catalog #: 470180-344
Supplier: ENTOMOLOGICAL RESEARCH
Specifications
- Description:Unsexed Adults, Pack of 10
- Cat. no.:470180-344
- Supplier No.:WARD470180-344
Specifications
About this item
Cockroaches are widely used in labs for physiological, behavioral, structural, and developmental studies.
- Specimens come from cultures bred in the lab environment
This species is also used regularly to test the effectiveness of pesticides.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought