Live Paramecium aurelia
Catalog #: 470180-150
Supplier: Avantor
Specifications
- Description:Paramecium aurelia Culture for 100 Students
- Cat. no.:470180-150
- Supplier No.:WARD470180-150
Specifications
About this item
Paramecium aurelia is the smallest species of Paramecium we offer. Paramecium aurelia is characterized by cilia, which it uses for locomotion and feeing. Contains a macronucleus and two micronuclei. Grows in Dilute Alfalfa Medium (Hay Medium).
Customers Who Bought This Also Bought