Live Paramecium aurelia
Catalog #: 470180-148
Supplier: CONNECTICUT VALLEY BIO SUPPLY
Specifications
- Description:Paramecium aurelia (Small)
- No. of Students:30
- Cat. no.:470180-148
- Packaging:Tube
- Media:Hay culture medium
- Supplier No.:WARD470180-148
Specifications
About this item
Paramecium aurelia is the smallest species of Paramecium we offer. Paramecium aurelia is characterized by cilia, which it uses for locomotion and feeing. Contains a macronucleus and two micronuclei. Grows in Dilute Alfalfa Medium (Hay Medium).
Customers Who Bought This Also Bought