ContactUsLinkComponent
PORPHYRIDIUM 35 STUDENTS
Porphyridium
Catalog #: 470179-906
Supplier:  Ward's Science
PORPHYRIDIUM 35 STUDENTS
Porphyridium
Catalog #: 470179-906
Supplier:  Ward's Science
Specifications
  • Description:
    Porphyridium (Small)
  • No. of Students:
    30
  • Cat. no.:
    470179-906
  • Packaging:
    Tube
  • Media:
    Erdschreiber's medium
  • Supplier No.:
    WARD470179-906
Specifications
About this item
Marine. Unicellular. Erdschreiber’s medium.
Characterized by reddish plastids (rhodoplasts).
Customers Who Bought This Also Bought