Ward's® Live GFP producing E. coli Culture
Catalog #: 470179-088
Supplier: Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:E. coli Strain B21
- Storage Conditions:Refrigerator
- Cat. no.:470179-088
- Packaging:Plate
- Media:Luria agar with ampicillin
- Supplier No.:WARD470179-088
- Incubation Conditions:37 °C
Specifications
About this item
Eschericia coli culture that produces GFP, visible in the presence of UV light.
Ordering information: These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Caution: Although this organism is non pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Customers Who Bought This Also Bought